missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 3 Step-Assembly with Studded Bearings

Product Code. 11492370 Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep 24™
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11492370

Brand: MP Biomedicals™ 116002513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Replacement part for FastPrep-24™

With the FastPrep-24 Instrument, adapter and optimized Lysing Matrices, consistent reproducible results are obtained.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Tri-step Assembly with Bearing
For Use With (Equipment) FastPrep 24™
Packaging Quantity 1 Ea.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.