missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MOZ Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566566

Product Code. 15929264

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SSAPQEQYGEC GEKSEATQEQ YTESEEQLVA SEEQPSQDGK PDLPKRRLSE GVEPWRGQLK KSPEALKCRL TEGSE Highest antigen sequence identity to the following orthologs - mouse 67%, rat 61%.

Histone acetyltransferase that acetylates lysine residues in histone H3 and histone H4 (in vitro). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity. May act as a transcriptional coactivator for RUNX1 and RUNX2. Acetylates p53/TP53 at 'Lys-120' and 'Lys-382' and controls its transcriptional activity via association with PML.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MOZ
Polyclonal
Unconjugated
Kat6a
1500036M03; 9930021N24Rik; histone acetyltransferase KAT6A; histone acetyltransferase MYST3; K(lysine) acetyltransferase 6A; KAT6A; lysine acetyltransferase 6A; monocytic leukemia zinc finger homolog; Monocytic leukemia zinc finger protein; Moz; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3; MRD32; MYST histone acetyltransferase (monocytic leukemia) 3; Myst3; MYST-3; runt-related transcription factor binding protein 2; Runt-related transcription factor-binding protein 2; RUNXBP2; ZC2HC6A; Zfp220; zinc finger protein 220; ZNF220
Rabbit
Antigen affinity chromatography
RUO
7994
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q92794
Kat6a
Recombinant Human MOZ. Recombinant protein control fragment (Product #RP-107223).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.