missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568046
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Histone acetyltransferase that acetylates lysine residues in histone H3 and histone H4 (in vitro). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity. May act as a transcriptional coactivator for RUNX1 and RUNX2. Acetylates p53/TP53 at 'Lys-120' and 'Lys-382' and controls its transcriptional activity via association with PML.
Specifications
| MOZ | |
| Polyclonal | |
| Unconjugated | |
| Kat6a | |
| 1500036M03; 9930021N24Rik; histone acetyltransferase KAT6A; histone acetyltransferase MYST3; K(lysine) acetyltransferase 6A; KAT6A; lysine acetyltransferase 6A; monocytic leukemia zinc finger homolog; Monocytic leukemia zinc finger protein; Moz; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3; MRD32; MYST histone acetyltransferase (monocytic leukemia) 3; Myst3; MYST-3; runt-related transcription factor binding protein 2; Runt-related transcription factor-binding protein 2; RUNXBP2; ZC2HC6A; Zfp220; zinc finger protein 220; ZNF220 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 244349, 306571, 7994 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5TKR9, Q8BZ21, Q92794 | |
| Kat6a | |
| Synthesized peptide derived from human MOZ. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction