missing translation for 'onlineSavingsMsg'
Learn More

Moxonidine, MedChemExpress

Product Code. 16458266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458266 Supplier Medchem Express Supplier No. HYB0374
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Moxonidine(BDF5895) is a selective agonist at the imidazoline receptor subtype 1, used as antihypertensive agent.

Moxonidine(BDF5895) is a selective agonist at the imidazoline receptor subtype 1, used as antihypertensive agent.

Specifications

Chemical Name or Material Moxonidine
Physical Form Powder
CAS 75438-57-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H12ClN5O
For Use With (Application) Neuroscience-Neuromodulation
Synonym BDF5895
Solubility Information DMSO : 20 mg/mL (82.75 mM; Need ultrasonic)
SMILES CC1=NC(OC)=C(NC2=NCCN2)C(Cl)=N1
Molecular Weight (g/mol) 241.68
Formula Weight 241.68
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.