missing translation for 'onlineSavingsMsg'
Learn More

Moxisylyte hydrochloride, MedChemExpress

Product Code. p-200101239
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 16412349

Brand: Medchem Express HYB1435

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Moxisylyte (hydrochloride) is (alpha 1-blocker) antagonist, it can vasodilates cerebral vessels without reducing blood pressure.

Moxisylyte (hydrochloride) is (alpha 1-blocker) antagonist, it can vasodilates cerebral vessels without reducing blood pressure.

Specifications

Chemical Name or Material Moxisylyte hydrochloride
CAS 964-52-3
Color White
Purity Grade Notes Research
Molecular Formula C16H26ClNO3
Quantity 1 g
Synonym Thymoxamine hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (158.31 mM) ∣DMSO : 20 mg/mL (63.32 mM; Need ultrasonic)
SMILES CC(C1=C(OCCN(C)C)C=C(C)C(OC(C)=O)=C1)C.[H]Cl
Molecular Weight (g/mol) 315.84
Formula Weight 315.84
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.