missing translation for 'onlineSavingsMsg'
Learn More

Moxidectin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30315429
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315429 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315429 Supplier Dr. Ehrenstorfer Supplier No. DREA15335000AL100
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Moxidectin
CAS 113507-06-5
Analyte or Component Names Moxidectin
Shipping Condition Room Temperature
Molecular Formula C37 H53 N O8
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Moxidectin, (2aE,2'R,4E,4'E,5'S,6R,6'S,8E,11R,15S,17aR,20R,20aR,20bS)-6'-[(1E)-1,3-Dimethylbut-1-enyl]-20,20b-dihydroxy-4'-(methoxyimino)-5',6,8,19-tetramethyl-3',4',5',6,6',7,10,11,14,15,17a,20,20a,20b-tetradecahydrospiro[2H,17H-11,15-methanofuro[4,3,2-pq][2,6]benzodioxacyclooctadecine-13,2'-pyran]-17-one, (6R,23E,25S)-5O-demethyl-28-deoxy-25-[(1E)-1,3-dimethylbut-1-enyl]-6,28-epoxy-23-(methoxyimino)milbemycin B, Milbemycin B, 5-O-demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-6,28-epoxy-23-(methoxyimino)-, (6R,23E,25S)-, Milbemycin B, 5-O-demethyl-28-deoxy-25-(1,3-dimethyl-1-butenyl)-6,28-epoxy-23-(methoxyimino)-, [6R,23E,25S(E)]-, Milbemycin B, 5-O-demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-butenyl]-6,28-epoxy-23-(methoxyimino)-, (6R,23E,25S)- (9CI), (6R,23E,25S)-5-O-Demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-6,28-epoxy-23-(methoxyimino)milbemycin B, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran], milbemycin B deriv., 23-Methoxime-LL-F 28249α, CL 301423, Cydectin, Equest, ProHeart 6, Quest, Quest (parasiticide), Vetdectin Oral Drench
InChI Formula InChI=1S/C37H53NO8/c1-21(2)14-25(6)33-26(7)31(38-42-8)19-36(46-33)18-29-17-28(45-36)13-12-23(4)15-22(3)10-9-11-27-20-43-34-32(39)24(5)16-30(35(40)44-29)37(27,34)41/h9-12,14,16,21-22,26,28-30,32-34,39,41H,13,15,17-20H2,1-8H3/b10-9+,23-12+,25-14+,27-11+,38-31+/t22-,26-,28+,29-,30-,32+,33+,34+,36-,37+/m0/s1
SMILES CO\N=C\1/C[C@]2(C[C@@H]3C[C@@H](C\C=C(/C)\C[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O3)[C@]45O)C)O2)O[C@@H]([C@H]1C)\C(=C\C(C)C)\C
IUPAC Name (2aE,2'R,4E,4'E,5'S,6R,6'S,8E,11R,15S,17aR,20R,20aR,20bS)-6'-[(1E)-1,3-Dimethylbut-1-enyl]-20,20b-dihydroxy-4'-(methoxyimino)-5',6,8,19-tetramethyl-3',4',5',6,6',7,10,11,14,15,17a,20,20a,20b-tetradecahydrospiro[2H,17H-11,15-methanofuro[4,3,2-pq][2,6]benzodioxacyclooctadecine-13,2'-pyran]-17-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 639.82
Quantity 1 mL
Formula Weight 639.3771 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.