missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mover Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | Mover |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256242
|
Novus Biologicals
NBP2-55810 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685697
|
Novus Biologicals
NBP2-55810-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mover Polyclonal specifically detects Mover in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Mover | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FAM79A, family with sequence similarity 79, member A, FLJ21811, h-mover, Mossy fiber terminal-associated vertebrate-specific presynaptic protein, MOVER, Protein FAM79A, RP11-46F15.3, tumor protein p63 regulated 1-like, tumor protein p63-regulated gene 1-like protein | |
| TPRG1L | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 127262 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PLRQTLWPLSIHDPTRRARVKEYFVFRPGSIEQAVEEIRVVVRPVEDGEIQGVWLLTEVDHWNNEK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title