Learn More
MOV10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10370-1-AP-150UL
 View more versions of this product
Product Code. 16856713
  • £349.00 / 150µL
Description
Probable RNA helicase. Required for RNA-mediated gene silencing by the RNA-induced silencing complex (RISC). Required for both miRNA-mediated translational repression and miRNA- mediated cleavage of complementary mRNAs by RISC. Also required for RNA-directed transcription and replication of the human hepatitis delta virus (HDV). Interacts with small capped HDV RNAs derived from genomic hairpin structures that mark the initiation sites of RNA-dependent HDV RNA transcription.
Specifications
Specifications
MOV10
Polyclonal
Unconjugated
MOV10
fSAP113, gb110, KIAA1631, MOV10, Putative helicase MOV 10
Rabbit
Antigen Affinity Chromatography
RUO
17454, 310756, 4343
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.55 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23249, Q9HCE1
Mov10
MOV10 Fusion Protein Ag0384
150 μL
Primary
Human, Mouse, Rat