missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOV10 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-77314AF647
This item is not returnable.
View return policy
Description
MOV10 Polyclonal antibody specifically detects MOV10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MOV10 | |
| Polyclonal | |
| Western Blot | |
| DKFZp667O1423, EC 3.6.1, EC 3.6.4.13, FLJ32791, KIAA1631mouse) homolog, Mov10, Moloney leukemia virus 10, homolog (mouse) | |
| The immunogen recognized by this antibody maps to a region between residue 953 and 1003 of human Moloney leukemia virus 10, homolog using the numbering given in entry NP_066014.1 (GeneID 4343). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4343 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction