missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ZIP4H/Tex11 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5627
This item is not returnable.
View return policy
Description
ZIP4H/Tex11 Polyclonal specifically detects ZIP4H/Tex11 in Mouse samples. It is validated for Western Blot.
Specifications
| ZIP4H/Tex11 | |
| Polyclonal | |
| Unconjugated | |
| Q14AT2 | |
| TEX11 | |
| E. coli-derived recombinant mouse ZIP4H/Tex11 Met768-Leu947 Accession # Q14AT2 | |
| 100 μg | |
| Primary | |
| Detects mouse ZIP4H/Tex11 in Western blots. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| testis expressed 11, testis expressed sequence 11, testis-expressed sequence 11 protein, Tex11, TGC1, TSGA3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 56159 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction