missing translation for 'onlineSavingsMsg'
Learn More

Mouse YM1/Chitinase 3-like 3 Biotinylated Antibody, R&D Systems™

Product Code. 18080882
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18080882 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18080882 Supplier R&D Systems Supplier No. BAF2446

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 6 publications

YM1/Chitinase 3-like 3 Polyclonal specifically detects YM1/Chitinase 3-like 3 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YM1/Chitinase 3-like 3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O35744
Gene Alias AI505981, Chi3l3, Chitinase 3 like 3, Chitinase 3-like 3, ECF-L, Ym1
Gene Symbols Chil3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse YM1/Chitinase 3-like 3 Tyr22-Tyr398 Accession # O35744
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1117
Test Specificity Detects mouse YM1/Chitinase 3-like 3 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.