missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse YM1/Chitinase 3-like 3 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB2446
This item is not returnable.
View return policy
Description
YM1/Chitinase 3-like 3 Monoclonal specifically detects YM1/Chitinase 3-like 3 in Mouse samples. It is validated for Western Blot.
Specifications
| YM1/Chitinase 3-like 3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| O35744 | |
| Chil3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse YM1/Chitinase 3-like Tyr22-Tyr398 Accession # O35744 | |
| 100 μg | |
| Primary | |
| Detects mouse YM1/Chitinase 3-like in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 281926 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AI505981, Chi3l3, Chitinase 3 like 3, Chitinase 3-like 3, ECF-L, Ym1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1117 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction