missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse XL IL-10, Luminex™ Performance Assay

Product Code. 18402440
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402440 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402440 Supplier R&D Systems™ Supplier No. LMXL417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Species Mouse
Gene ID (Entrez) 3586
Synonym CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.