missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse XL FGF basic/FGF2/bFGF, Luminex™ Performance Assay
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Mouse |
| Gene ID (Entrez) | 2247 |
| Synonym | Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?