missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse XL CCL2/JE/MCP-1, Luminex™ Performance Assay
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Kit
missing translation for 'unitSize'
1 set
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Mouse |
| Gene ID (Entrez) | 6347 |
| Synonym | C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction