missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Wnt-2b Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF3900
This item is not returnable.
View return policy
Description
Wnt-2b Polyclonal specifically detects Wnt-2b in Mouse samples. It is validated for Western Blot.
Specifications
| Wnt-2b | |
| Polyclonal | |
| Biotin | |
| O70283 | |
| WNT2B | |
| E. coli-derived recombinant mouse Wnt-2b Leu64-Glu104 and Arg251-Ala312 Accession # O70283 | |
| 50 μg | |
| Primary | |
| Detects mouse Wnt-2b in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) Wnt-2 is observed and less than 5% cross-reactivity with rhWnt-7a, rhWnt-9a, recombinant mouse (rm) Wnt-1, rmWnt-3a, rmWnt-4, rmWnt-5a, rmWnt-5b, rmWnt-8a, rmWnt-8b, rmWnt-9b, rmWnt-10a, rmWnt-10b, rmWnt-11, and rmWnt-16 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Protein Wnt-13, protein Wnt-2b, wingless-type MMTV integration site family, member 2B, Wnt-13, WNT13XWNT2, Xenopus, homolog of, Wnt2b, XWNT2, XWNT2wingless-type MMTV integration site family, member 13 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7482 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction