missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Wnt-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1620-SP
This item is not returnable.
View return policy
Description
Wnt-1 Polyclonal specifically detects Wnt-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Wnt-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Int-1, INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1), Wnt1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7471 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q3UR96 | |
| WNT1 | |
| E. coli-derived recombinant mouse Wnt-1 peptide Ser52-Glu92 and Met234-Arg313 Accession # Q3UR96 | |
| 25 μg | |
| Primary | |
| Detects mouse Wnt-1 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Wnt-3a and rmWnt-5a is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction