missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VSIG4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4674-SP
This item is not returnable.
View return policy
Description
VSIG4 Polyclonal specifically detects VSIG4 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| VSIG4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11326 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| NP_808457 | |
| VSIG4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse VSIG4 His20-Pro187 Accession # NP_808457 | |
| 25 μg | |
| Primary | |
| Detects mouse VSIG4 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) VSIG4 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction