missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VSIG2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4766
This item is not returnable.
View return policy
Description
VSIG2 Polyclonal specifically detects VSIG2 in Mouse samples. It is validated for Western Blot.
Specifications
| VSIG2 | |
| Polyclonal | |
| Unconjugated | |
| Q9Z109 | |
| VSIG2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse VSIG2 Val25-Ala244 (Glu240Asp) Accession # Q9Z109 | |
| 100 μg | |
| Primary | |
| Detects mouse VSIG2 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) VSIG2 is observed and less than 10% cross-reactivity with rhVSIG3 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| 2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23584 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction