missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Vitronectin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3875-SP
This item is not returnable.
View return policy
Description
Vitronectin Monoclonal specifically detects Vitronectin in Mouse samples. It is validated for Western Blot.
Specifications
| Vitronectin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAH12690 | |
| VTN | |
| Mouse myeloma cell line NS0-derived recombinant mouse Vitronectin Asp20-Lys478 Accession # AAH12690 | |
| 25 μg | |
| Primary | |
| Detects mouse Vitronectin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Vitronectin is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 347312 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT, VTN | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7448 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction