missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Vitronectin Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB3875
Additional Details : Weight : 0.00970kg
Description
Vitronectin Monoclonal specifically detects Vitronectin in Mouse samples. It is validated for Western Blot.Specifications
Vitronectin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
AAH12690 | |
VTN | |
Mouse myeloma cell line NS0-derived recombinant mouse Vitronectin Asp20-Lys478 Accession # AAH12690 | |
100 μg | |
Primary | |
Detects mouse Vitronectin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Vitronectin is observed. | |
Mouse | |
Purified |
Western Blot | |
347312 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT, VTN | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7448 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
For Research Use Only