missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Mouse Vegfa (120 amino acids) Recombinant Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Sequence: APTTEGEQKSHEVIKFMDVYQRSYCRPIETLVDIFQEYPDEIEYIFKPSCVPLMRCAGCCNDEALECVPTSESNITMQIMRIKPHQSQHIGEMSFLQHSRCECRPKKDRTKPEKCDKRPRR
Specifications
Specifications
| For Use With (Application) | Functional Study, SDS-PAGE |
| Formulation | Lyophilized |
| Gene ID (Entrez) | 22339 |
| Molecular Weight (g/mol) | 28.2kDa |
| Name | Vegfa (Mouse) Recombinant Protein |
| Preparation Method | Escherichia coli expression system |
| Quantity | 10 μg |
| Immunogen | MAPTTEGEQKSHEVIKFMDVYQRSYCRPIETLVDIFQEYPDEIEYIFKPSCVPLMRCAGCCNDEALECVPTSESNITMQIMRIKPHQSQHIGEMSFLQHSRCECRPKKDRTKPEKCDKPRR |
| Storage Requirements | Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction