missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse VEGF Quantikine ELISA Kit

Product Code. p-4753980
Click to view available options
Quantity:
1 Kit (1 Plate)
6 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18752074

Brand: R&D Systems™ SMMV00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse VEGF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse VEGF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse VEGF in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 3pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity 164 and 120 amino acid residue forms of Mouse VEGF
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 22339
Quantity 6 Plates
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.