missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VEGF-D Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF469
This item is not returnable.
View return policy
Description
VEGF-D Polyclonal specifically detects VEGF-D in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VEGF-D | |
| Polyclonal | |
| Unconjugated | |
| P97946 | |
| FIGF | |
| E. coli-derived recombinant mouse VEGF-D Phe98-Ser206 Accession # P97946 | |
| 100 μg | |
| Primary | |
| Detects mouse VEGF-D in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant human (rh) VEGF-D is observed, and less than 1% cross-reactivity with recombinant mouse (rm) VEGF 115, rmVEGF 120, rhVEGF, rhVEGF-B, and rmVEGF 165 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| c-fos induced growth factor (vascular endothelial growth factor D), FIGF, vascular endothelial growth factor D, VEGFD, VEGF-DVEGFDc-Fos-induced growth factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2277 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction