missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse VEGF-C ELISA Kit (Colorimetric)

Product Code. 18039263 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18039263 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18039263 Supplier Novus Biologicals™ Supplier No. NBP278893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This kit recognizes Mouse VEGF-C in samples.

No Significant cross-reactivity or interference between Mouse VEGF-C and analogues was observed.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P97953
Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 7424
Gene Symbol VEGFC
Host Species Mouse
Quantity 1 Kit
Research Discipline Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target VEGF-C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.