missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VEGF-B186 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB767-SP
This item is not returnable.
View return policy
Description
VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot.
Specifications
| VEGF-B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7423 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 124112 | |
| Western Blot 1 ug/mL | |
| P49766.2 | |
| Vegfa | |
| E. coli-derived recombinant mouse VEGF-B 186 Pro22-Ala207 Accession # P49766.2 | |
| 25 μg | |
| Primary | |
| Detects mouse VEGF-B 186 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) VEGF 165 rhVEGF-B 186 recombinant mouse (rm) VEGF-B 167 rhVEGF-C, rmVEGF-D, rhCTGF, rhPlGF, or rhLDGF is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction