Learn More
Invitrogen™ Mouse VEGF-A ELISA Kit
Description
Includes
Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to mouse VEGF-A
Biotin-Conjugate anti-mouse VEGF-A polyclonal antibody
Streptavidin-HRP
Mouse VEGF-A Standard lyophilized, 4ng/mL upon reconstitution
Sample Diluent
Assay Diluent
Conjugate Diluent Concentrate 20x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
The Mouse Vascular Endothelial cell Growth Factor A (Ms VEGF-A) ELISA quantitates Ms VEGF-A in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms VEGF-A. Principle of the method The Mouse VEGF-A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | Q00731 |
| Assay Range | 62.5-4000 pg/mL |
| Assay Sensitivity | 20.0 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.