Learn More
Mouse VE-Statin Isoform 1 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand:  R&D Systems BAF3089
Product Code. 18179210
  • £457.00 / 50µg
Description
VE-Statin/EGFL7 Polyclonal specifically detects VE-Statin/EGFL7 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VE-Statin/EGFL7
Polyclonal
Biotin
Q9QXT5
EGFL7
E. coli-derived recombinant mouse VE-Statin Isoform 1 Thr21-Leu275 Accession # Q9QXT5
50 μg
Primary
Detects mouse VE-Statin Isoform 1 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human VE-Statin is observed.
Mouse
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
EGFL7, EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VEStatin, VE-STATIN, Zneu1
Goat
Affinity Purified
RUO
51162
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.