missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse VCAM-1/CD106 Quantikine ELISA Kit
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
Description
The Mouse sVCAM-1/CD106 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse VCAM-1/CD106 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul).
Specifications
Specifications
| Assay Range | 0.31 - 20ng/mL |
| Assay Sensitivity | 0.06ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Serum |
| Specificity | Natural and Recombinant Mouse sVCAM-1 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 22329 |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction