missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VCAM-1/CD106 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£452.00 - £1780.00
Specifications
| Antigen | VCAM-1/CD106 |
|---|---|
| Clone | 112734 |
| Dilution | Western Blot 2 ug/mL, Simple Western 100 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Flow Cytometry, CyTOF, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18749678
![]() |
R&D Systems
MAB6432-100 |
100 μg |
£452.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701089
![]() |
R&D Systems
MAB6432-500 |
500 μg |
£1780.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| VCAM-1/CD106 | |
| Western Blot 2 ug/mL, Simple Western 100 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| VCAM1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 112734 | |
| Western Blot, Flow Cytometry, CyTOF, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P29533 | |
| 7412 | |
| Mouse myeloma cell line NS0-derived recombinant mouse VCAM-1 Phe25-Glu698 (predicted) Accession # P29533 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant mouse (rm) VCAM-1 in Western blots and ELISAs. In sandwich immunoassays, no cross-reactivity or interference was observed with recombinant human VCAM-1, rmICAM-1, rmE-Selectin, rmL-Selectin or rmP-Selectin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title