missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Urotensin-II R Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB9245-SP
This item is not returnable.
View return policy
Description
Urotensin-II R/GPR14 Monoclonal specifically detects Urotensin-II R/GPR14 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Urotensin-II R/GPR14 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2837 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Flow Cytometry, CyTOF | |
| 443707 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UrotensinIIR, Urotensin-IIR, UTR, UTR2, UTS2R | |
| Mouse Urotensin-II R synthetic peptide | |
| 25 μg | |
| Primary | |
| Detects mouse Urotensin-II R. Stains mouse Urotensin-II R transfectants but not irrelevant transfectants in flow cytometry. | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction