missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse uPA/Urokinase (NP_032899.1) VersaClone cDNA

Product Code. 18701857
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18701857

Brand: R&D Systems™ RDC3775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene u-Plasminogen Activator/Urokinase
Accession Number NP_032899.1
Gene ID (Entrez) 5328
Gene Alias ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.