missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Universal Protein Whole Tissue Lysate (Adult Whole Normal)

Product Code. 18237623 Shop All Bio Techne Products
Click to view available options
Quantity:
4 Vials
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 18237623

Brand: Novus Biologicals™ NB82089208

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Mouse Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from mouse. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Universal
Quantity 4 Vials
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Universal Protein Lysate: Mouse Normal Tissues
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Mouse
Host Species Mouse
Concentration 5 mg/mL
Product Type Mouse Universal Protein Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.