missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse UGRP1/SCGB3A2 Antibody, R&D Systems™

Goat Polyclonal Antibody
£186.00 - £506.00
Specifications
| Antigen | UGRP1/SCGB3A2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748017
![]() |
R&D Systems
AF3465-SP |
25 μg |
£186.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18021582
|
R&D Systems
AF3465 |
100 μg |
£506.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UGRP1/SCGB3A2 Polyclonal specifically detects UGRP1/SCGB3A2 in Mouse samples. It is validated for Western Blot.Specifications
| UGRP1/SCGB3A2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| LU103, Pneumo secretory protein 1, PnSP-1, PNSP1secretoglobin family 3A member 2, SCGB3A2, secretoglobin, family 3A, member 2, UGRP1pnSP-1, Uteroglobin-related protein 1 | |
| SCGB3A2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q920H1 | |
| 117156 | |
| E. coli-derived recombinant mouse UGRP1/SCGB3A2 isoform C Leu22-Leu139 Accession # Q920H1 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse UGRP1/SCGB3A2 isoforms C and A in direct ELISAs and Western blots. It is also expected to recognize isoform B in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human UGRP1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title