missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Tyro3/Dtk Alexa Fluor 647-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB759R-100UG
This item is not returnable.
View return policy
Description
Dtk/TYRO3 Monoclonal specifically detects Dtk/TYRO3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Dtk/TYRO3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P55144 | |
| TYRO3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Dtk Ala31-Ser418 Accession # P55144 | |
| RUO | |
| 7301 | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| 109646 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Dtk extracellular domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Dtk, recombinant mouse (rm) Mer, or rmAxl is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction