missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse TWEAK/TNFSF12 DuoSet ELISA
Click to view available options
Conjugate:
Biotin
Unit Size:
1 set
Description
The Mouse TWEAK/TNFSF12 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse TWEAK/TNFSF12.
Specifications
Specifications
| Conjugate | Biotin |
| Assay Range | 15.60 to 1000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 21944 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction