missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TWEAK R/TNFRSF12 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1610
This item is not returnable.
View return policy
Description
TWEAK R/TNFRSF12 Polyclonal specifically detects TWEAK R/TNFRSF12 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| TWEAK R/TNFRSF12 | |
| Polyclonal | |
| Unconjugated | |
| Q9CR75 | |
| TNFRSF12A | |
| E. coli-derived recombinant mouse TWEAK R/TNFRSF12 Glu28-Trp79 Accession # Q9CR75 | |
| 100 μg | |
| Primary | |
| Detects mouse TWEAK R in ELISAs and Western blots. In sandwich ELISAs, less than 2% cross-reactivity with recombinant human TWEAK R is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, Fn14, FN14tumor necrosis factor receptor superfamily member 12A, TNFRSF12, TNFRSF12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 51330 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction