missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TWEAK R/TNFRSF12 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB16101T-100UG
This item is not returnable.
View return policy
Description
TWEAK R/TNFRSF12 Monoclonal specifically detects TWEAK R/TNFRSF12 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| TWEAK R/TNFRSF12 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9CR75.1 | |
| TNFRSF12A | |
| E. coli-derived recombinant mouse TWEAK R Glu28-Trp79 Accession # Q9CR75.1 | |
| RUO | |
| 51330 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 314636 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, Fn14, FN14tumor necrosis factor receptor superfamily member 12A, TNFRSF12, TNFRSF12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse TWEAK R/TNFRSF12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) EDA, rmNeogenin, recombinant human TWEAK R, rmOPG, or rmTNF R1 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction