missing translation for 'onlineSavingsMsg'
Learn More

Mouse TSLP Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18797368
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18797368 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18797368 Supplier R&D Systems Supplier No. AF555SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

TSLP Polyclonal specifically detects TSLP in Mouse samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSLP
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.1-0.4 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9JIE6
Gene Alias thymic stromal lymphopoietin
Gene Symbols TSLP
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TSLP Tyr20-Glu140 Accession # Q9JIE6
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85480
Test Specificity Detects mouse TSLP in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.