missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Tryptase epsilon/BSSP-4 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2059
This item is not returnable.
View return policy
Description
Tryptase epsilon/BSSP-4/PRSS22 Monoclonal specifically detects Tryptase epsilon/BSSP-4/PRSS22 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Tryptase epsilon/BSSP-4/PRSS22 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BSP-2, BSSP4, BSSP-4, hBSSP-4, MGC9599, prosemin, protease, serine S1 family member 22, protease, serine, 22, PRSS22, serine protease 22, serine protease 26, SP001LA, tryptase epsilon, UNQ302/PRO343 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64063 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunoprecipitation | |
| 246744 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q7TML0 | |
| PRSS22 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tryptase epsilon/BSSP-4 Ala33-Ser306 (predicted) Accession # Q7TML0 | |
| 500 μg | |
| Primary | |
| Detects mouse Tryptase epsilon/BSSP-4 in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction