missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Tryptase beta-1/MCP-7/Mcpt7 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1937
This item is not returnable.
View return policy
Description
Tryptase beta-1/MCP-7/Mcpt7 Polyclonal specifically detects Tryptase beta-1/MCP-7/Mcpt7 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Tryptase beta-1/MCP-7/Mcpt7 | |
| Polyclonal | |
| Unconjugated | |
| Q02844 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 17230 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Tpsab1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tryptase beta-1/MCP-7/Mcpt7 Ile29-Phe273 Accession # Q02844 | |
| 100 μg | |
| Primary | |
| Detects mouse Tryptase beta-1/MCP-7/Mcpt7 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) TPSB2 is observed, 5% cross-reactivity with rhTPS1 is observed, and less than 1% cross-reactivity with rhTPSG1 and recombinant mouse MCPT1 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction