missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TROP-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
£167.00 - £539.00
Specifications
| Antigen | TROP-2 |
|---|---|
| Clone | 168215 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745499
![]() |
R&D Systems
MAB1122-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120881
|
R&D Systems
MAB1122 |
500 μg |
£539.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TROP-2 Monoclonal specifically detects TROP-2 in Mouse samples. It is validated for Western Blot.Specifications
| TROP-2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8BGV3 | |
| 4070 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TROP-2 Ser82-Thr268 Accession # Q8BGV3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TROP-2 in direct ELISAs and Western blots. |
| 168215 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, gp50, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, T16, TACSTD2, TROP2, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| TACSTD2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title