missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TORC3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB6840-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TORC3/CRTC3 Monoclonal specifically detects TORC3/CRTC3 in Mouse samples. It is validated for Western Blot.
Spezifikation
| TORC3/CRTC3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q91X84 | |
| Crtc3 | |
| E. coli-derived recombinant mouse TORC3 Leu153-Pro275 Accession # Q91X84 | |
| 25 μg | |
| Primary | |
| Detects mouse TORC3 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human TORC3 and no cross-reactivity with recombinant mouse (rm) TORC1 or rmTORC2 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 643729 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CREB regulated transcription coactivator 3, CRTC3, FLJ21868, TORC-3, TORC3CREB-regulated transcription coactivator 3, Transducer of CREB protein 3, transducer of regulated cAMP response element-binding protein (CREB) 3, Transducer of regulated cAMP response element-binding protein 3, transducer of regulated CREB protein 3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64784 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur