missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TORC2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB6458-SP
This item is not returnable.
View return policy
Description
TORC2 Monoclonal specifically detects TORC2 in Mouse samples. It is validated for Western Blot.
Specifications
| TORC2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q3U182 | |
| CRTC2 | |
| E. coli-derived recombinant mouse TORC2 Lys454-Ser612 Accession # Q3U182 | |
| 25 μg | |
| Primary | |
| Detects mouse TORC2 in Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 632307 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CREB regulated transcription coactivator 2, CREB-regulated transcription coactivator 2, CRTC2, TORC-2, TORC2transducer of regulated cAMP response element-binding protein (CREB) 2, Transducer of CREB protein 2, Transducer of regulated cAMP response element-binding protein 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 200186 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction