missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TNFRH3/TNFRSF26 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB5330G-100UG
This item is not returnable.
View return policy
Description
TNFRH3/TNFRSF26 Monoclonal specifically detects TNFRH3/TNFRSF26 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| TNFRH3/TNFRSF26 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P83626 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse TNFRH3/TNFRSF26 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Fas, rmTNF RI, and rmTNF RII is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 509027 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Tnfrsf26 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TNFRH3/TNFRSF26 Ser20-Lys162 Accession # P83626 | |
| RUO | |
| 8718 | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction