missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TNF RII/TNFRSF1B Antibody, R&D Systems™
Hamster Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB426
This item is not returnable.
View return policy
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| TNF RII/TNFRSF1B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TNFRSF1B | |
| E. coli-derived recombinant mouse TNF RII/TNFRSF1B Extracellular domain | |
| 500 μg | |
| Primary | |
| Detects mouse TNF RII/TNFRSF1B in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) TNF RII, recombinant mouse (rm) TNF RI, rhTNF RI, rm4-1BB, or rmOPG is observed. In Western blots, approximately 25% cross-reactivity with rhDR3 and rhTNF RII is observed and no cross-reactivity with rm4-1BB, rmCD27, rmCD30, rmCD40, rmFas, rmGITR, rhHVEM, rhNGF R, rmOPG, rmRANK, rmTNF RI, rmTROY, or rmEDAR is observed. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF, ELISA | |
| TR7554 | |
| Western Blot 1 ug/mL, Flow Cytometry 1 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| Hamster | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7133 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction