missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TNF RI/TNFRSF1A Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems BAF425
This item is not returnable.
View return policy
Description
TNF RI/TNFRSF1A Polyclonal specifically detects TNF RI/TNFRSF1A in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| TNF RI/TNFRSF1A | |
| Polyclonal | |
| Biotin | |
| P25118 | |
| TNFRSF1A | |
| E. coli-derived recombinant mouse TNF RI/TNFRSF1A Ile22-Ala212 Accession # P25118 | |
| 50 μg | |
| Primary | |
| Detects mouse TNF RI/TNFRSF1A in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with recombinant human (rh) TNF-alpha , recombinant rat TNF-alpha , recombinant porcine TNF-alpha , rhTNF-beta , recombinant mouse TNF sRII, rhTNF sRII, and rhTNF sRI is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7132 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction