missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse TNF alpha, Recombinant Protein

Product Code. p-7092381
Click to view available options
Quantity:
10 μg
5 μg
Unit Size:
5µg
Each
This item is not returnable. View return policy

Product Code. 15587516

Brand: Invitrogen™ 14832162

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Mouse TNF-a is a 17 kDa factor produced by macrophages, monocytes, neutrophils, CD4+ T cells and NK cells. A 26 kDa form of TNF-α is expressed as a membrane-bound molecule. TNF-α is cytolytic and plays an important role in immune regulation. Dimers and trimers of TNF-α have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_013693
For Use With (Application) Cytokine Bioassay
Formulation Sterile liquid; 10mM sodium phosphate, pH 7.2, 150mM NaCl, 0.5% BSA. 0.22μm filtered.
Molecular Weight (g/mol) The protein is not methionylated at the N-terminal and has a predicted molecular mass of 17,258. The DTT-reduced and non-reduced protein migrate as 17kDa polypeptides on SDS-PAGE.
Name TNF alpha
Purification Method SDS-PAGE
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Structural Form E. coli expressed amino acids Leu 80-Leu 235 of mouse TNF- alpha accession # NM_013693
Endotoxin Concentration Less than 0.01ng/μg cytokine as determined by the LAL assay.
Gene Symbol Tumor Necrosis Factor alpha, TNFa TNF-a
Biological Activity The ED50 of this protein, as measured by L929 cytotoxicity assay, is less than or equal to 25pg/mL. This corresponds to a specific activity of greater than or equal to 4 x 107 Units/mg.
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Purity or Quality Grade >98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.