missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse TNF-alpha Quantikine ELISA Kit
Click to view available options
Quantity:
1 Plate
2 Plates
6 Plates
Unit Size:
1 set
Description
The Mouse TNF-alpha Quantikine ELISA Kit quantifies mouse TNF-alpha in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications
Specifications
| Assay Range | 10.90 - 700pg/mL |
| Assay Sensitivity | 7.21pg/mL |
| Conjugate | HRP |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Mouse TNF-alpha |
| Sufficient For | 2 x 96-well Plates |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 21926 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction