missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TNF-alpha Quantikine ELISA Kit

Product Code. 18732454
Click to view available options
Quantity:
1 Plate
2 Plates
6 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18732454

Brand: R&D Systems™ SMTA00B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse TNF-alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse TNF-alpha Quantikine ELISA Kit quantifies mouse TNF-alpha in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY

Specifikationer

Assay Range 10.90 - 700pg/mL
Assay Sensitivity 7.21pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Mouse TNF-alpha
Sufficient For 6 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 21926
Quantity 6 Plates
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.